Gene Transcripts

Gene Symbol
Wdr93
Entrez Gene ID
626359
Gene Name
WD repeat domain 93
Gene Aliases
EG626359
Chromosome Location
Chr. 7: 79743135 - 79785952 on Build GRCm38
UniGene ID
Mm.387216
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037927.1 NP_001033016.1 2 257
GenBank mRNA AB205021.1 - 2 257
BC150920.1 - 1 227 9087831

View Full Product Details