Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010.2 | NP_001001.2 | 5 | 639 | |
GenBank mRNA | AB062123.1 | - | 5 | 625 | |
AK093634.1 | - | 1840 | |||
AK291517.1 | - | 5 | 638 | ||
AK311861.1 | - | 5 | 636 | ||
BC000524.2 | - | 4 | 601 | 2961137 | |
BC009427.2 | - | 4 | 592 | ||
BC013296.2 | - | 5 | 622 | 3030319 | |
BC027620.1 | - | 5 | 642 | 4698843 | |
BC035447.1 | - | 3 | 463 | ||
BC071907.1 | - | 5 | 627 | 4900361 | |
BC071908.1 | - | 5 | 627 | 5089456 | |
BC094826.1 | - | 5 | 623 | 5481378 | |
J03537.1 | - | 4 | 597 |