Gene Transcripts

Gene Symbol
Vmn2r56
Entrez Gene ID
629079
Gene Name
vomeronasal 2, receptor 56
Gene Aliases
EG629079
Chromosome Location
Chr. 7: 12693860 - 12733320 on Build GRCm38
UniGene ID
Mm.469869
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104648.1 NP_001098118.1 1 74
GenBank mRNA AB919109.1 - 1 74

View Full Product Details