Gene Transcripts

Gene Symbol
Dcpp2
Entrez Gene ID
630537
Gene Name
demilune cell and parotid protein 2
Gene Aliases
Dcpp-2, EG630537, SPT-2
Chromosome Location
Chr. 17: 23898722 - 23900787 on Build GRCm38
UniGene ID
Mm.436677
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039238.2 NP_001034327.2 4 335
GenBank mRNA BC059096.1 - 3 354
BC171996.1 - 336 9054282
DQ237937.1 - 335

View Full Product Details