Gene Transcripts

Gene Symbol
Dmrtc1b
Entrez Gene ID
632708
Gene Name
DMRT-like family C1b
Gene Aliases
Dmrt8.2, Gm714
Chromosome Location
Chr. X: 102707881 - 102715354 on Build GRCm38
UniGene ID
Mm.297715
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039116.2 NP_001034205.2 5 417
GenBank mRNA BC150829.1 - 5 417
DQ316775.1 - 4 394
DQ316776.1 - 4 554
DQ316777.1 - 5 472

View Full Product Details