Gene Transcripts

Gene Symbol
Vmn2r16
Entrez Gene ID
384220
Gene Name
vomeronasal 2, receptor 16
Gene Aliases
EG384220
Chromosome Location
Chr. 5: 109330381 - 109364481 on Build GRCm38
UniGene ID
Mm.469880
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104627.1 NP_001098097.1 4 1482

View Full Product Details