Gene Transcripts

Gene Symbol
Vmn2r85
Entrez Gene ID
623734
Gene Name
vomeronasal 2, receptor 85
Gene Aliases
EG623734
Chromosome Location
Chr. 10: 130418261 - 130429612 on Build GRCm38
UniGene ID
Mm.469840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102602.1 NP_001096072.1 3 1040

View Full Product Details