Gene Transcripts

Gene Symbol
Trim63
Entrez Gene ID
433766
Gene Name
tripartite motif-containing 63
Gene Aliases
MuRF1, RF1, Rnf28
Chromosome Location
Chr. 4: 134315088 - 134329629 on Build GRCm38
UniGene ID
Mm.331961
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC147663.1 - 7 1185
BC147666.1 - 7 1185 9087455

View Full Product Details