Gene Transcripts
- Gene Symbol
- Fsip1
- Entrez Gene ID
- 71313
- Gene Name
- fibrous sheath-interacting protein 1
- Gene Aliases
- 1700012M13Rik, 4933432K11Rik
- Chromosome Location
- Chr. 2: 118130261 - 118256987 on Build GRCm38
- UniGene ID
- Mm.85046
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF448787.1 |
- |
10 |
1310 |
|
BC125024.1 |
- |
10 |
1202 |
40056533 |
View Full Product Details