Gene Transcripts

Gene Symbol
ATXN7
Entrez Gene ID
6314
Gene Name
ataxin 7
Gene Aliases
ADCAII, OPCA3, SCA7
Chromosome Location
Chr. 3: 63864557 - 64003462 on Build GRCh38
UniGene ID
Hs.476595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000333.3 NP_000324.1 3, 4 886
NM_001177387.1 NP_001170858.1 2, 3 443
GenBank mRNA AF032103.1 - 1, 2 25
AF032105.1 - 3, 4 886
AJ000517.1 - 3, 4 894

View Full Product Details