Gene Transcripts

Gene Symbol
Vmn2r91
Entrez Gene ID
665210
Gene Name
vomeronasal 2, receptor 91
Gene Aliases
EG665210
Chromosome Location
Chr. 17: 18085057 - 18136643 on Build GRCm38
UniGene ID
Mm.469864
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104540.1 NP_001098010.1 1 193

View Full Product Details