Gene Transcripts

Gene Symbol
Vmn2r5
Entrez Gene ID
667060
Gene Name
vomeronasal 2, receptor 5
Gene Aliases
EG667060
Chromosome Location
Chr. 3: 64490783 - 64534962 on Build GRCm38
UniGene ID
Mm.484957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104618.1 NP_001098088.1 5 1895

View Full Product Details