Gene Transcripts

Gene Symbol
Vmn2r21
Entrez Gene ID
546912
Gene Name
vomeronasal 2, receptor 21
Gene Aliases
EG546912
Chromosome Location
Chr. 6: 123492508 - 123533406 on Build GRCm38
UniGene ID
Mm.469876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104635.1 NP_001098105.1 1 200

Gene Symbol
Vmn2r20
Entrez Gene ID
667180
Gene Name
vomeronasal 2, receptor 20
Gene Aliases
EG667180
Chromosome Location
Chr. 6: 123385262 - 123418061 on Build GRCm38
UniGene ID
Mm.469877
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104634.1 NP_001098104.1 1 200

View Full Product Details