Gene Transcripts

Gene Symbol
Ighv4-2
Entrez Gene ID
668423
Gene Name
immunoglobulin heavy variable V4-2
Gene Aliases
Chromosome Location
Chr. 12: 114013144 - 114013439 on Build GRCm38
UniGene ID
Mm.425752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA Z72454.1 - 56

View Full Product Details