Gene Transcripts
- Gene Symbol
- SEMG1
- Entrez Gene ID
- 6406
- Gene Name
- semenogelin I
- Gene Aliases
- CT103, SEMG, SGI, dJ172H20.2
- Chromosome Location
- Chr. 20: 45206964 - 45209773 on Build GRCh38
- UniGene ID
- Hs.1968
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003007.4 |
NP_002998.1
|
2 |
1292 |
|
GenBank mRNA |
J04440.1 |
- |
2 |
1208 |
|
View Full Product Details