Gene Transcripts

Gene Symbol
Ugt1a9
Entrez Gene ID
394434
Gene Name
UDP glucuronosyltransferase 1 family, polypeptide A9
Gene Aliases
UDPGT, UDPGT 1-9, UGT1-09, UGT1.9, UGTP4, Ugt1a12, mUGTBr/P
Chromosome Location
Chr. 1: 88070779 - 88220002 on Build GRCm38
UniGene ID
Mm.300095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201644.2 NP_964006.2 1 311
GenBank mRNA AY227200.1 - 1 290
BC019434.1 - 1 285
BC138699.1 - 1 290 8861721
BC146021.1 - 1 290 40131324
L27122.1 - 1 236
S57479.1 - 1 236

Gene Symbol
Ugt1a10
Entrez Gene ID
394430
Gene Name
UDP glycosyltransferase 1 family, polypeptide A10
Gene Aliases
A13, Ugt1a13
Chromosome Location
Chr. 1: 88055411 - 88220002 on Build GRCm38
UniGene ID
Mm.300095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201641.2 NP_964003.2 1 337
GenBank mRNA AY227201.1 - 1 296
BC089569.1 - 1 337 30120494

View Full Product Details