Gene Transcripts

Gene Symbol
Myc
Entrez Gene ID
24577
Gene Name
myelocytomatosis oncogene
Gene Aliases
RNCMYC, c-myc, mMyc
Chromosome Location
Chr. 7: 102586313 - 102591240 on Build Rnor_6.0
UniGene ID
Rn.12072
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012603.2 NP_036735.2 3 1690
GenBank mRNA AY679729.1 - 3 1527
AY679730.1 - 3 1690
BC091699.1 - 3 1496 7303974

View Full Product Details