Gene Transcripts

Gene Symbol
Pdgfb
Entrez Gene ID
24628
Gene Name
platelet-derived growth factor beta polypeptide
Gene Aliases
SIS, c-sis
Chromosome Location
Chr. 7: 121215458 - 121233092 on Build Rnor_6.0
UniGene ID
Rn.198230
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031524.1 NP_113712.1 6 1064
GenBank mRNA CB783426.1 - 1 58
Z14117.1 - 6 645

View Full Product Details