Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017050.1 | NP_058746.1 | 2 | 235 | |
GenBank mRNA | BC082800.1 | - | 2 | 215 | |
FQ209495.1 | - | 2 | 236 | ||
FQ209941.1 | - | 2 | 231 | ||
FQ220715.1 | - | 2 | 235 | ||
M21060.1 | - | 2 | 138 | ||
M25157.1 | - | 2 | 194 | ||
X05634.1 | - | 2 | 134 | ||
Y00404.1 | - | 2 | 235 |