Gene Transcripts
- Gene Symbol
- Tf
- Entrez Gene ID
- 24825
- Gene Name
- transferrin
- Gene Aliases
- Tfn, Trf
- Chromosome Location
- Chr. 8: 111694570 - 111721275 on Build Rnor_6.0
- UniGene ID
- Rn.91296
- Species
- Rattus norvegicus
- Gene Symbol
- Srprb
- Entrez Gene ID
- 300965
- Gene Name
- signal recognition particle receptor, B subunit
- Gene Aliases
- Ab2-417, Ba1-667, Cc1-8
- Chromosome Location
- Chr. 8: 111673479 - 111687426 on Build Rnor_6.0
- UniGene ID
- Rn.138078
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY325214.1 |
- |
|
937 |
|
AY325230.1 |
- |
|
937 |
|
AY325241.1 |
- |
|
937 |
|
View Full Product Details