Gene Transcripts

Gene Symbol
Tnp1
Entrez Gene ID
24839
Gene Name
transition protein 1
Gene Aliases
Chromosome Location
Chr. 9: 80294837 - 80295446 on Build Rnor_6.0
UniGene ID
Rn.9959
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017056.2 NP_058752.1 1 69
GenBank mRNA BC078850.1 - 1 69
M17096.1 - 1 70

View Full Product Details