Gene Transcripts

Gene Symbol
Vcsa1
Entrez Gene ID
24867
Gene Name
variable coding sequence A1
Gene Aliases
Arp, P2-VA1, RATSMR1A, Smr1, Smr1g
Chromosome Location
Chr. 14: 21490217 - 21709084 on Build Rnor_6.0
UniGene ID
Rn.169239
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012684.4 NP_036816.4 3 249
GenBank mRNA A07543.1 - 3 256
CB769246.1 - 3 261
M63112.1 - 3 256

View Full Product Details