Gene Transcripts

Gene Symbol
Rnf112
Entrez Gene ID
24916
Gene Name
ring finger protein 112
Gene Aliases
BFP, Bfb, Zfp179, Znf179
Chromosome Location
Chr. 10: 47719034 - 47726950 on Build Rnor_6.0
UniGene ID
Rn.7544
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138613.1 NP_619516.1 3 393
XM_006246441.2 XP_006246503.1 5 1505
XM_006246445.2 XP_006246507.1 3 396
XM_008767774.1 XP_008765996.1 5 1933
XM_008767775.1 XP_008765997.1 5 1933
XM_008767776.1 XP_008765998.1 5 1642
XM_008767777.1 XP_008765999.1 4 578
GenBank mRNA AF054586.1 - 3 393
FQ212014.1 - 2 309

View Full Product Details