Gene Transcripts

Gene Symbol
SIX3
Entrez Gene ID
6496
Gene Name
SIX homeobox 3
Gene Aliases
HPE2
Chromosome Location
Chr. 2: 44941898 - 44946077 on Build GRCh38
UniGene ID
Hs.567336
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005413.3 NP_005404.1 1 442
GenBank mRNA AJ012611.1 - 1 442
AL162671.1 - 1 423
BC030289.1 - 1 266

View Full Product Details