Gene Transcripts

Gene Symbol
Vars
Entrez Gene ID
25009
Gene Name
valyl-tRNA synthetase
Gene Aliases
Bat6, G7a, TrsVal, Vars2
Chromosome Location
Chr. 20: 4993537 - 5008259 on Build Rnor_6.0
UniGene ID
Rn.12314
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053292.1 NP_445744.1 28 3492
XM_006256047.2 XP_006256109.1 28 3708
GenBank mRNA BC099236.1 - 8 897
FQ216008.1 - 3 479

View Full Product Details