Gene Transcripts
- Gene Symbol
- Scgb2a1
- Entrez Gene ID
- 25010
- Gene Name
- secretoglobin, family 2A, member 1
- Gene Aliases
- Psbp1
- Chromosome Location
- Chr. 1: 225825356 - 225830355 on Build Rnor_6.0
- UniGene ID
- Rn.103180
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080770.3 |
|
2 |
131 |
|
- Gene Symbol
- Scgb2a2
- Entrez Gene ID
- 361725
- Gene Name
- secretoglobin, family 2A, member 2
- Gene Aliases
- Chromosome Location
- Chr. 1: 225899014 - 225902676 on Build Rnor_6.0
- UniGene ID
- Rn.103180
- Species
- Rattus norvegicus
View Full Product Details