Gene Transcripts
- Gene Symbol
- Cryba1
- Entrez Gene ID
- 25583
- Gene Name
- crystallin, beta A1
- Gene Aliases
- BA3A1C, beta-A3
- Chromosome Location
- Chr. 10: 65161152 - 65167504 on Build Rnor_6.0
- UniGene ID
- Rn.20326
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013056.1 |
NP_037188.1
|
6 |
578 |
|
GenBank mRNA |
X15143.1 |
- |
4 |
459 |
|
View Full Product Details