Gene Transcripts

Gene Symbol
Tnni3
Entrez Gene ID
29248
Gene Name
troponin I type 3 (cardiac)
Gene Aliases
TnI, cTNI
Chromosome Location
Chr. 1: 72882806 - 72886490 on Build Rnor_6.0
UniGene ID
Rn.64141
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017144.2 NP_058840.1 8 694
GenBank mRNA DQ062462.1 - 6 590
M57679.1 - 8 677
M92074.1 - 7 644
X58499.1 - 8 695

View Full Product Details