Gene Transcripts

Gene Symbol
Map1b
Entrez Gene ID
29456
Gene Name
microtubule-associated protein 1B
Gene Aliases
Mtap1b
Chromosome Location
Chr. 2: 29675391 - 29768750 on Build Rnor_6.0
UniGene ID
Rn.220177
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019217.1 NP_062090.1 5 3522
XM_008760661.1 XP_008758883.1 4 3187
GenBank mRNA X60370.1 - 3 3070

View Full Product Details