Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003801.1 | NP_001003801.1 | 11 | 1290 | |
NM_001003802.1 | NP_001003802.1 | 12 | 1279 | ||
NM_003078.3 | NP_003069.2 | 12 | 1318 | ||
XM_011516521.2 | XP_011514823.1 | 11 | 1397 | ||
XM_017012556.1 | XP_016868045.1 | 11 | 1133 | ||
GenBank mRNA | AK127595.1 | - | 12 | 1826 | |
AY450430.1 | - | 11 | 1594 | ||
AY450431.1 | - | 12 | 1279 | ||
BC002628.2 | - | 12 | 1279 | 3608512 | |
BC110350.1 | - | 11 | 1209 | 5212363 | |
BX648385.1 | - | 10 | 2041 | ||
U66619.1 | - | 12 | 1327 |