Gene Transcripts

Gene Symbol
Atp2b4
Entrez Gene ID
29600
Gene Name
ATPase, Ca++ transporting, plasma membrane 4
Gene Aliases
PMCA4
Chromosome Location
Chr. 13: 50091639 - 50153808 on Build Rnor_6.0
UniGene ID
Rn.9986
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005871.1 NP_001005871.1 11 2026
XM_008769446.1 XP_008767668.1 12 2409
XM_008769447.1 XP_008767669.1 12 3046
XM_008769448.1 XP_008767670.1 11 2373
XM_008769449.1 XP_008767671.1 12 2410
XM_008769450.1 XP_008767672.1 11 2374
GenBank mRNA U15408.1 - 11 2026

View Full Product Details