Gene Transcripts

Gene Symbol
Rgs11
Entrez Gene ID
54291
Gene Name
regulator of G-protein signaling 11
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.84083
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019338.1 NP_062211.1 486
GenBank mRNA CB605941.1 - 368

View Full Product Details