Gene Transcripts
- Gene Symbol
- Pdcd2
- Entrez Gene ID
- 58934
- Gene Name
- programmed cell death 2
- Gene Aliases
- Chromosome Location
- Chr. 1: 57512968 - 57518458 on Build Rnor_6.0
- UniGene ID
- Rn.6959
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031638.1 |
NP_113826.1
|
4, 5 |
815 |
|
GenBank mRNA |
M80601.1 |
- |
4, 5 |
597 |
|
View Full Product Details