Gene Transcripts

Gene Symbol
Nxph4
Entrez Gene ID
59316
Gene Name
neurexophilin 4
Gene Aliases
Nph4
Chromosome Location
Chr. 7: 70833662 - 70842405 on Build Rnor_6.0
UniGene ID
Rn.42882
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021680.2 NP_067712.2 2 717
XM_008765418.1 XP_008763640.1 2 713
GenBank mRNA AF042714.1 - 2 788
BC081805.1 - 2 717 7108207

View Full Product Details