Gene Transcripts

Gene Symbol
SP3
Entrez Gene ID
6670
Gene Name
Sp3 transcription factor
Gene Aliases
SPR2
Chromosome Location
Chr. 2: 173906459 - 173965702 on Build GRCh38
UniGene ID
Hs.531587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017371.4 NP_001017371.3 2 1213
NM_001172712.1 NP_001166183.1 4 1555
NM_003111.4 NP_003102.1 4 1564
GenBank mRNA AB209334.1 - 1 677
AK304199.1 - 4 1136
AY070137.1 - 2 1213
AY441957.1 - 4 1564
AY441958.1 - 2 903
BC126414.1 - 4 1086 8992130
BC143928.1 - 4 1077 9052446
M97191.2 - 2 824
X68560.1 - 2 782

View Full Product Details