Gene Transcripts

Gene Symbol
Slc18a3
Entrez Gene ID
60422
Gene Name
solute carrier family 18 (vesicular acetylcholine transporter), member 3
Gene Aliases
VACht, rVAT
Chromosome Location
Chr. 16: 8682668 - 8685529 on Build Rnor_6.0
UniGene ID
Rn.9987
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031663.2 NP_113851.1 1 1757
GenBank mRNA U09211.1 - 1 1757
U09838.1 - 1 1014

View Full Product Details