Gene Transcripts

Gene Symbol
Treh
Entrez Gene ID
60576
Gene Name
trehalase (brush-border membrane glycoprotein)
Gene Aliases
Chromosome Location
Chr. 8: 48983802 - 48998072 on Build Rnor_6.0
UniGene ID
Rn.163015
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136141.1 NP_001129613.1 7, 8 757
XM_006242956.2 XP_006243018.1 6, 7 629
GenBank mRNA AF038043.1 - 6, 7 678

View Full Product Details