Gene Transcripts

Gene Symbol
SPN
Entrez Gene ID
6693
Gene Name
sialophorin
Gene Aliases
CD43, GALGP, GPL115, LSN
Chromosome Location
Chr. 16: 29662950 - 29670507 on Build GRCh38
UniGene ID
Hs.632188
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030288.2 NP_001025459.1 2 567
NM_003123.4 NP_003114.1 2 534
GenBank mRNA AK292626.1 - 2 506
AK313750.1 - 2 506
BC012350.1 - 2 454 4564347
DB108930.1 - 2 534
J04536.1 - 2 494
X60502.1 - 2 538

View Full Product Details