Gene Transcripts

Gene Symbol
AMPH
Entrez Gene ID
273
Gene Name
amphiphysin
Gene Aliases
AMPH1
Chromosome Location
Chr. 7: 38383694 - 38631567 on Build GRCh38
UniGene ID
Hs.592182
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001635.3 NP_001626.1 6 643
NM_139316.2 NP_647477.1 6 643
XM_006715689.3 XP_006715752.1 6 590
XM_006715690.3 XP_006715753.1 6 590
XM_006715691.3 XP_006715754.1 6 590
XM_011515271.2 XP_011513573.1 6 590
XM_017011995.1 XP_016867484.1 6 590
XM_017011996.1 XP_016867485.1 6 590
GenBank mRNA AF034996.1 - 6 496
AF498097.1 - 6 427
AK291447.1 - 6 447
BC034376.1 - 6 434 5181100
U07616.1 - 6 537
X81438.1 - 6 501

View Full Product Details