Gene Transcripts

Gene Symbol
SPRR2B
Entrez Gene ID
6701
Gene Name
small proline rich protein 2B
Gene Aliases
Chromosome Location
Chr. 1: 153070226 - 153071608 on Build GRCh38
UniGene ID
Hs.568239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017418.2 NP_001017418.1 2 431
XM_017002174.1 XP_016857663.1 2 427
GenBank mRNA BF090370.1 - 1 303

View Full Product Details