Gene Transcripts

Gene Symbol
SPRR3
Entrez Gene ID
6707
Gene Name
small proline rich protein 3
Gene Aliases
Chromosome Location
Chr. 1: 153001747 - 153003856 on Build GRCh38
UniGene ID
Hs.139322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097589.1 NP_001091058.1 2 467
NM_005416.2 NP_005407.1 3 551
GenBank mRNA AJ243667.1 - 3 425
AK311823.1 - 2 465
AY118269.1 - 2 443
BC017802.1 - 3 441 4288753
DA763011.1 - 2 465
EF553525.1 - 3 1491

View Full Product Details