Gene Transcripts
- Gene Symbol
- Nrgn
- Entrez Gene ID
- 64356
- Gene Name
- neurogranin
- Gene Aliases
- RC3
- Chromosome Location
- Chr. 8: 40015049 - 40023193 on Build Rnor_6.0
- UniGene ID
- Rn.11236
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024140.2 |
NP_077054.1
|
4 |
1333 |
|
GenBank mRNA |
L09119.1 |
- |
4 |
1333 |
|
View Full Product Details