Gene Transcripts

Gene Symbol
Prkab2
Entrez Gene ID
64562
Gene Name
protein kinase, AMP-activated, beta 2 non-catalytic subunit
Gene Aliases
Chromosome Location
Chr. 2: 199831990 - 199847623 on Build Rnor_6.0
UniGene ID
Rn.207202
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022627.2 NP_072149.1 6 734
GenBank mRNA AF182717.1 - 6 736

View Full Product Details