Gene Transcripts

Gene Symbol
TRIM21
Entrez Gene ID
6737
Gene Name
tripartite motif containing 21
Gene Aliases
RNF81, RO52, Ro/SSA, SSA, SSA1
Chromosome Location
Chr. 11: 4384897 - 4393696 on Build GRCh38
UniGene ID
Hs.532357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003141.3 NP_003132.2 6 907
GenBank mRNA AK225564.1 - 4 620
BC010861.1 - 6 878 3900540
M34551.1 - 5 832
M62800.1 - 6 879

View Full Product Details