Gene Transcripts

Gene Symbol
Arf5
Entrez Gene ID
79117
Gene Name
ADP-ribosylation factor 5
Gene Aliases
Chromosome Location
Chr. 4: 55715744 - 55718670 on Build Rnor_6.0
UniGene ID
Rn.28244
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024149.1 NP_077063.1 2, 3 251
GenBank mRNA BC087692.1 - 2, 3 213
L12384.1 - 2, 3 251

View Full Product Details