Gene Transcripts

Gene Symbol
HSPA13
Entrez Gene ID
6782
Gene Name
heat shock protein family A (Hsp70) member 13
Gene Aliases
STCH
Chromosome Location
Chr. 21: 14371115 - 14383188 on Build GRCh38
UniGene ID
Hs.352341
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006948.4 NP_008879.3 2 166
GenBank mRNA AK128540.1 - 2 124
AK223282.1 - 2 124
AK299004.1 - 2 115
AK312396.1 - 2 121
BC036370.2 - 2 128 4837136
BI548162.1 - 2 192
U04735.1 - 2 133

View Full Product Details