Gene Transcripts

Gene Symbol
Tmsb4x
Entrez Gene ID
81814
Gene Name
thymosin beta 4, X-linked
Gene Aliases
Chromosome Location
Chr. X: 28593405 - 28617267 on Build Rnor_6.0
UniGene ID
Rn.201192
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031136.1 NP_112398.1 3 544
GenBank mRNA BC058137.1 - 3 557
FQ211950.1 - 3 554
FQ229728.1 - 3 553
M34043.1 - 3 544

View Full Product Details