Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003162.3 | NP_003153.2 | 7 | 842 | |
XM_005264519.4 | XP_005264576.1 | 7 | 977 | ||
XM_011533073.2 | XP_011531375.1 | 7 | 977 | ||
GenBank mRNA | AJ223814.1 | - | 7 | 842 | |
AK026934.1 | - | 4 | 413 | ||
AK307836.1 | - | 3 | 519 | ||
BC106878.2 | - | 8 | 759 | ||
BC106879.2 | - | 8 | 805 | 40027865 |