Gene Transcripts

Gene Symbol
Dynlt1
Entrez Gene ID
83462
Gene Name
dynein light chain Tctex-type 1
Gene Aliases
AGS2, Tctex1
Chromosome Location
Chr. 1: 47206892 - 47213782 on Build Rnor_6.0
UniGene ID
Rn.29901
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031318.1 NP_112608.1 3, 4 200
XM_006227901.2 XP_006227963.1 3, 4 265
GenBank mRNA AB010119.1 - 3, 4 189
AJ131437.1 - 3, 4 200
BC166879.1 - 3, 4 232 6919674

View Full Product Details