Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001055.3 | NP_001046.2 | 8 | 1104 | |
NM_177529.2 | NP_803565.1 | 8 | 1135 | ||
NM_177530.2 | NP_803566.1 | 8 | 1260 | ||
NM_177534.2 | NP_803878.1 | 7 | 1459 | ||
NM_177536.3 | NP_803880.1 | 6 | 1141 | ||
XM_017023604.1 | XP_016879093.1 | 9 | 1207 | ||
XM_017023605.1 | XP_016879094.1 | 9 | 1204 | ||
XM_017023606.1 | XP_016879095.1 | 9 | 1189 | ||
XM_017023607.1 | XP_016879096.1 | 11 | 1750 | ||
XM_017023608.1 | XP_016879097.1 | 8 | 1122 | ||
XM_017023609.1 | XP_016879098.1 | 8 | 1172 | ||
XM_017023610.1 | XP_016879099.1 | 8 | 1203 | ||
XM_017023611.1 | XP_016879100.1 | 10 | 1677 | ||
XM_017023612.1 | XP_016879101.1 | 11 | 1601 | ||
XM_017023613.1 | XP_016879102.1 | 9 | 1416 | ||
GenBank mRNA | AB062428.1 | - | 7 | 1042 | |
AB209149.1 | - | 7819 | |||
BC000923.2 | - | 8 | 1054 | 3447900 | |
BC110887.1 | - | 11 | 1517 | 5190558 | |
L10819.1 | - | 8 | 1024 | ||
L19955.1 | - | 7 | 1027 | ||
L19999.1 | - | 8 | 1055 | ||
U09031.1 | - | 1027 | |||
U26309.1 | - | 7 | 1050 | ||
X78283.1 | - | 8 | 1145 |